Assay Details
Gene Symbol: LOC494127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036691.1 | 1 | 150 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC440173 | LOC440173 | -41.8kb | Sense | Chr. 9 - 87008451 - 87042126 on NCBI Build GRCh38 | |
Downstream | chromosome 9 open reading frame 170 | C9orf170 | 63.4kb | Anti-sense | Chr. 9 - 87148644 - 87159726 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001297.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137135 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137060 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797207 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137088 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137126 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797241 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797202 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137078 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137099 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797299 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797205 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16235927 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137103 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137138 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797231 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137064 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137061 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797206 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797242 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797224 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797217 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137139 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16235931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797317 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797289 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137059 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797248 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797266 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797318 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137066 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797255 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797209 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137056 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797237 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137132 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797257 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797323 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137069 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137053 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137141 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137076 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797321 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137137 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797208 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137083 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137131 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137062 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797221 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137085 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27137039 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06797211 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8162190 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162188 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162189 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8162187 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09016279.hg.1 | TC09001297.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09016279.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010639.hg.1 | ENST00000437925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172636.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172636.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172636.hg.1 | TC0900010639.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172637.hg.1 | ENST00000437925.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172637.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172637.hg.1 | ENST00000437925.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172637.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172637.hg.1 | TC0900010639.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172638.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172638.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172638.hg.1 | TC0900010639.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010639.hg.1 | ENST00000437925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010639.hg.1 | NR_036691.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037656 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037656 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 714115 | - | - |
