Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011409.hg.1 |
ENST00000606367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011409.hg.1 |
ENST00000606367.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198101.hg.1 |
TC0600014101.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011409.hg.1 |
ENST00000606367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011409.hg.1 |
ENST00000606367.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000421350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000449934.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
ENST00000616296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_000247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001177519.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014101.hg.1 |
NM_001289154.1 |
Refseq |