Assay Details
Gene Symbol: MUC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198815.1 | NP_001185744.1 | 293 | 69 | ||
NM_001318484.1 | NP_001305413.1 | 258 | 69 | |||
NM_001322469.1 | NP_001309398.1 | 258 | 69 | |||
GenBank mRNA | AB560770.1 | - | 293 | 69 | ||
AB600271.1 | - | 307 | 69 | |||
AB600272.1 | - | 258 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000354.hg.1, TC6_apd_hap1000032.hg.1, TC6_cox_hap2000052.hg.1, TC6_dbb_hap3000045.hg.1, TC6_mann_hap4000045.hg.1, TC6_mcf_hap5000039.hg.1, TC6_qbl_hap6000045.hg.1, TC6_ssto_hap7000040.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3214914 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3408232 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000596.hg.1 | TC6_ssto_hap7000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000596.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000743.hg.1 | TC6_mann_hap4000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000743.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000745.hg.1 | TC6_mann_hap4000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000745.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000780.hg.1 | TC6_qbl_hap6000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000780.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000781.hg.1 | TC6_qbl_hap6000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000781.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000594.hg.1 | TC6_ssto_hap7000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000594.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000349.hg.1 | TC6_apd_hap1000032.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000349.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000350.hg.1 | TC6_apd_hap1000032.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000350.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000351.hg.1 | TC6_apd_hap1000032.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000351.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000771.hg.1 | TC6_dbb_hap3000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000771.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003384.hg.1 | TC06000354.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003384.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000788.hg.1 | TC6_cox_hap2000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000788.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000770.hg.1 | TC6_dbb_hap3000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000770.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000742.hg.1 | TC6_mann_hap4000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000742.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000783.hg.1 | TC6_qbl_hap6000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000783.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000790.hg.1 | TC6_cox_hap2000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000790.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000791.hg.1 | TC6_cox_hap2000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000791.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000643.hg.1 | TC6_mcf_hap5000039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000643.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000644.hg.1 | TC6_mcf_hap5000039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000644.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000782.hg.1 | TC6_qbl_hap6000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000782.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000593.hg.1 | TC6_ssto_hap7000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000593.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003381.hg.1 | TC06000354.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003381.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003382.hg.1 | TC06000354.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003382.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000789.hg.1 | TC6_cox_hap2000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000789.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000595.hg.1 | TC6_ssto_hap7000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000595.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003383.hg.1 | TC06000354.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003383.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000352.hg.1 | TC6_apd_hap1000032.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000352.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000769.hg.1 | TC6_dbb_hap3000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000769.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000772.hg.1 | TC6_dbb_hap3000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000772.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000744.hg.1 | TC6_mann_hap4000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000744.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000641.hg.1 | TC6_mcf_hap5000039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000641.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000642.hg.1 | TC6_mcf_hap5000039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000642.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007557.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151500.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151500.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151500.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151500.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151500.hg.1 | TC0600007557.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151498.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151498.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151498.hg.1 | TC0600007557.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151501.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151501.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151501.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151501.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151501.hg.1 | TC0600007557.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151502.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151502.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151502.hg.1 | ENST00000561890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151502.hg.1 | NM_001198815.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151502.hg.1 | TC0600007557.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031265 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031265 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1855975 | NM_001198815.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1855975 | AB560770.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1855975 | AB600271.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1855975 | AB600272.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1855975 | CV572205.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5670564 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_135148 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855975 | NM_001198815.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855975 | AB560770.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855975 | AB600271.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855975 | AB600272.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855975 | CV572205.1 | human_transcriptome |
