Assay Details
Gene Symbol: MCIDAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190787.1 | NP_001177716.1 | 3 - 4 | 489 | 94 | |
GenBank mRNA | FR854393.1 | - | 3 - 4 | 312 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001360.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
regulation of DNA replication
transcription, DNA-templated
cell cycle
regulation of mitotic cell cycle
cilium assembly
motile cilium assembly
positive regulation of transcription from RNA polymerase II promoter
centriole assembly
multi-ciliated epithelial cell differentiation
transcription coactivator activity
protein binding
identical protein binding
regulation of DNA replication
transcription, DNA-templated
cell cycle
regulation of mitotic cell cycle
cilium assembly
motile cilium assembly
positive regulation of transcription from RNA polymerase II promoter
centriole assembly
multi-ciliated epithelial cell differentiation
transcription coactivator activity
protein binding
identical protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05018949.hg.1 | TC05001360.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018949.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018949.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018949.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010758.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000513312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000513468.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | ENST00000515336.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | NM_001190787.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500175033.hg.1 | TC0500010758.hg.1 | Clariom D TC |
