Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
8166817 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002126.hg.1 |
TC0X000191.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002126.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007013.hg.1 |
ENST00000423387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007013.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007013.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147697.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147697.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147697.hg.1 |
TC0X00007013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147696.hg.1 |
ENST00000423387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147696.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147696.hg.1 |
ENST00000423387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147696.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147696.hg.1 |
TC0X00007013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007013.hg.1 |
ENST00000423387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007013.hg.1 |
NM_001195522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007013.hg.1 |
NM_001195522.2 |
Refseq |
Human Genome U95 Set |
66756_PM_at |
- |
- |
Human Genome U95 Set |
66756_at |
- |
- |