Assay Details
Gene Symbol: KCNH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204798.1 | NP_001191727.1 | 1 - 2 | 436 | 100 | |
NM_172057.2 | NP_742054.1 | 1 - 2 | 436 | 100 | ||
GenBank mRNA | AJ512214.1 | - | 1 - 2 | 127 | 100 | |
AJ609614.1 | - | 1 - 2 | 110 | 100 | ||
BQ890248.1 | - | 1 - 2 | 434 | 100 | ||
DQ120125.1 | - | 1 - 2 | 513 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
regulation of heart rate by hormone
signal transduction by protein phosphorylation
cellular response to drug
regulation of membrane potential
potassium ion homeostasis
cardiac muscle contraction
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
cardiac conduction
potassium ion export
potassium ion transmembrane transport
ventricular cardiac muscle cell action potential
membrane depolarization during action potential
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
potassium ion export across plasma membrane
membrane repolarization during ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transport
negative regulation of potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
negative regulation of potassium ion export
phosphorelay sensor kinase activity
inward rectifier potassium channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
protein binding
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
C3HC4-type RING finger domain binding
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
scaffold protein binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization
phosphorelay signal transduction system
regulation of heart rate by hormone
signal transduction by protein phosphorylation
cellular response to drug
regulation of membrane potential
potassium ion homeostasis
cardiac muscle contraction
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
cardiac conduction
potassium ion export
potassium ion transmembrane transport
ventricular cardiac muscle cell action potential
membrane depolarization during action potential
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
potassium ion export across plasma membrane
membrane repolarization during ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transport
negative regulation of potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
negative regulation of potassium ion export
phosphorelay sensor kinase activity
inward rectifier potassium channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
protein binding
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
C3HC4-type RING finger domain binding
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
scaffold protein binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000262186.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000330883.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000430723.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000461280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000473610.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | ENST00000532957.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_000238.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_001204798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172056.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013050.hg.1 | NM_172057.2 | Refseq |
