Assay Details
Gene Symbol: NT5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204813.1 | NP_001191742.1 | 5 - 6 | 1653 | 88 | |
NM_002526.3 | NP_002517.1 | 5 - 6 | 1653 | 88 | ||
GenBank mRNA | AK075008.1 | - | 5 - 6 | 1128 | 88 | |
AK075083.1 | - | 2 - 3 | 355 | 88 | ||
AK314661.1 | - | 5 - 6 | 1167 | 88 | ||
BC065937.1 | - | 5 - 6 | 1122 | 6163101 | 88 | |
DQ186653.1 | - | 5 - 6 | 1116 | 88 | ||
X55740.1 | - | 5 - 6 | 1146 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK075083.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
