Assay Details
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002443.3 | NP_002434.1 | 2 - 3 | 139 | 103 | |
GenBank mRNA | AK312108.1 | - | 2 - 3 | 139 | 103 | |
BC005257.1 | - | 2 - 3 | 139 | 3950847 | 103 | |
BM856408.1 | - | 2 - 3 | 150 | 103 | ||
BT006816.1 | - | 1 - 2 | 107 | 103 | ||
CR457015.1 | - | 1 - 2 | 107 | 103 | ||
M15885.1 | - | 1 - 2 | 118 | 103 | ||
S67815.1 | - | 2 - 3 | 127 | 103 | ||
U78976.1 | - | 1 - 2 | 131 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
