Assay Details
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006765.3 | NP_006756.2 | 2 - 3 | 645 | 94 | |
NM_178234.2 | NP_839952.1 | 2 - 3 | 645 | 94 | ||
XM_011544651.2 | XP_011542953.1 | 1 - 2 | 188 | 94 | ||
XM_011544652.2 | XP_011542954.1 | 1 - 2 | 188 | 94 | ||
XM_011544654.2 | XP_011542956.1 | 1 - 2 | 188 | 94 | ||
XM_017013860.1 | XP_016869349.1 | 1 - 2 | 188 | 94 | ||
XM_017013861.1 | XP_016869350.1 | 1 - 2 | 188 | 94 | ||
GenBank mRNA | BC010370.1 | - | 2 - 3 | 513 | 4334284 | 94 |
BT020002.1 | - | 2 - 3 | 301 | 94 | ||
U42349.1 | - | 2 - 3 | 458 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
