Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023011.3 | NP_075387.1 | 1 - 2 | 261 | 95 | |
NM_080687.2 | NP_542418.1 | 1 - 2 | 261 | 95 | ||
XM_011534844.1 | XP_011533146.1 | 1 - 2 | 239 | 95 | ||
XM_011534845.2 | XP_011533147.1 | 1 - 2 | 221 | 95 | ||
XM_011534846.1 | XP_011533148.1 | 1 - 2 | 208 | 95 | ||
XM_011534848.2 | XP_011533150.1 | 1 - 2 | 221 | 95 | ||
XM_017020709.1 | XP_016876198.1 | 1 - 2 | 221 | 95 | ||
XM_017020712.1 | XP_016876201.1 | 1 - 2 | 254 | 95 | ||
GenBank mRNA | AF318575.1 | - | 1 - 2 | 154 | 95 | |
AK294545.1 | - | 1 - 2 | 208 | 95 | ||
AK302003.1 | - | 1 - 2 | 197 | 95 | ||
AY013250.1 | - | 1 - 2 | 261 | 95 | ||
BC023569.1 | - | 1 - 2 | 225 | 4127465 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK302003.1, XM_011534845.2, XM_011534848.2, XM_017020709.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
