Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023011.3 | NP_075387.1 | 3 - 4 | 481 | 101 | |
XM_011534846.1 | XP_011533148.1 | 3 - 4 | 428 | 101 | ||
XM_011534848.2 | XP_011533150.1 | 3 - 4 | 966 | 101 | ||
GenBank mRNA | AF318575.1 | - | 3 - 4 | 374 | 101 | |
AK302003.1 | - | 3 - 4 | 417 | 101 | ||
AY013250.1 | - | 3 - 4 | 481 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK302003.1, XM_011534848.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
