Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023011.3 | NP_075387.1 | 8 - 9 | 1062 | 82 | |
NM_080687.2 | NP_542418.1 | 7 - 8 | 963 | 82 | ||
XM_005266202.4 | XP_005266259.1 | 5 - 6 | 688 | 82 | ||
XM_006719991.3 | XP_006720054.1 | 7 - 8 | 937 | 82 | ||
XM_011534844.1 | XP_011533146.1 | 6 - 7 | 830 | 82 | ||
XM_011534845.2 | XP_011533147.1 | 8 - 9 | 1590 | 82 | ||
XM_011534847.2 | XP_011533149.1 | 7 - 8 | 1402 | 82 | ||
XM_011534848.2 | XP_011533150.1 | 8 - 9 | 1547 | 82 | ||
XM_017020709.1 | XP_016876198.1 | 7 - 8 | 1774 | 82 | ||
XM_017020710.1 | XP_016876199.1 | 8 - 9 | 1159 | 82 | ||
XM_017020711.1 | XP_016876200.1 | 7 - 8 | 1060 | 82 | ||
GenBank mRNA | AF318575.1 | - | 8 - 9 | 955 | 82 | |
AK001069.1 | - | 3 - 4 | 339 | 82 | ||
AK097027.1 | - | 4 - 5 | 1024 | 82 | ||
AK302003.1 | - | 8 - 9 | 998 | 82 | ||
AY013250.1 | - | 8 - 9 | 1062 | 82 | ||
BC008694.1 | - | 4 - 5 | 475 | 82 | ||
BC023569.1 | - | 7 - 8 | 927 | 4127465 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC008694.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
