Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000581478.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
ENST00000582163.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_002443.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010516.hg.1 |
NM_138634.2 |
Refseq |