Assay Details
Gene Symbol: CDRT15L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190790.1 | NP_001177719.1 | 2 | 657 | 107 | |
GenBank mRNA | AI016778.1 | - | 1 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000289.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3356762 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 240889_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003563.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003563.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003563.hg.1 | TC17000289.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003564.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003564.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003564.hg.1 | TC17000289.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700151425.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151425.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151425.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151425.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151425.hg.1 | TC1700007230.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009803.hg.1 | ENST00000438438.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151426.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151426.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151426.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151426.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151426.hg.1 | TC1700007230.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | ENST00000447470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006992.hg.1 | NR_033865.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151427.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151427.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151427.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700151427.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700151427.hg.1 | TC1700007230.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000420162.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | ENST00000431716.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009830.hg.1 | NM_001007530.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | ENST00000399044.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007230.hg.1 | NM_001190790.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 240889_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240889_at | - | - |
Human Genome U95 Set | 78102_at | - | - |
Human Genome U95 Set | 78102_PM_at | - | - |
Human Genome U133 Set | 240889_PM_at | - | - |
Human Genome U133 Set | 240889_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009153 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009153 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1672275 | NM_001190790.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1672275 | AI016778.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672275 | NM_001190790.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672275 | AI016778.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672275 | NM_001190790.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672275 | AI016778.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672275 | NM_001190790.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672275 | AI016778.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672275 | NM_001190790.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672275 | AI016778.1 | human_transcriptome |
