Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008111.hg.1 |
ENST00000385278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008111.hg.1 |
NR_030381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008111.hg.1 |
ENST00000385278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008111.hg.1 |
NR_030381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155498.hg.1 |
ENST00000385278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155498.hg.1 |
NR_030381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155498.hg.1 |
ENST00000385278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155498.hg.1 |
NR_030381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155498.hg.1 |
TC0X00008111.hg.1 |
Clariom D TC |