Assay Details
| Gene Symbol: HLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006120.3 | NP_006111.2 | 2 | 432 | 93 | |
| GenBank mRNA | AK291858.1 | - | 2 | 368 | 93 | |
| BC011447.1 | - | 2 | 405 | 4245221 | 93 | |
| BC026279.1 | - | 2 | 361 | 4703588 | 93 | |
| CR450324.1 | - | 2 | 346 | 93 | ||
| X62744.1 | - | 2 | 391 | 93 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000374843.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395303.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000395305.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000456800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000475627.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000477541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | ENST00000480785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014277.hg.1 | NM_006120.3 | Refseq |

HLA-DMA
Genomic Map