Assay Details
| Gene Symbol: APCDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153000.4 | NP_694545.1 | 1 - 2 | 411 | 113 | |
| XM_011525603.1 | XP_011523905.1 | 1 - 2 | 404 | 113 | ||
| GenBank mRNA | AB056722.1 | - | 1 - 2 | 410 | 113 | |
| AB104887.1 | - | 1 - 2 | 410 | 113 | ||
| AK300743.1 | - | 2 - 3 | 213 | 113 | ||
| BC053324.1 | - | 2 - 3 | 382 | 6644230 | 113 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000051.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000553.hg.1 | ENST00000355285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000553.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000553.hg.1 | TC18000051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000554.hg.1 | ENST00000355285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000554.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000554.hg.1 | TC18000051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000552.hg.1 | ENST00000355285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000552.hg.1 | ENST00000423585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000552.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000552.hg.1 | TC18000051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000556.hg.1 | ENST00000355285.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000556.hg.1 | ENST00000423585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000556.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000556.hg.1 | TC18000051.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800145637.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000579685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | ENST00000584596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006675.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000578882.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145643.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145638.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145638.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145638.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145638.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145638.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | ENST00000582723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145642.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145639.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145639.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145639.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145639.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145639.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | NM_153000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | TC1800006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | ENST00000355285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | ENST00000423585.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800145640.hg.1 | NM_153000.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 131540 | - | - |

APCDD1
Genomic Map