Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145873.1 | NP_001139345.1 | 5 | 1161 | 86 | |
| NM_001768.6 | NP_001759.3 | 2 | 1019 | 86 | ||
| NM_171827.3 | NP_741969.1 | 2 | 1019 | 86 | ||
| NR_027353.1 | 3 | 605 | 86 | |||
| GenBank mRNA | AK097942.1 | - | 2 | 249 | 86 | |
| AK124156.1 | - | 5 | 1161 | 86 | ||
| AK300089.1 | - | 3 | 571 | 86 | ||
| AY039664.1 | - | 2 | 309 | 86 | ||
| BC025715.1 | - | 2 | 135 | 5227906 | 86 | |
| M12824.1 | - | 2 | 217 | 86 | ||
| M12828.1 | - | 2 | 195 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CD8A
Genomic Map