Assay Details
| Gene Symbol: LCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002298.4 | NP_002289.2 | 9 | 1206 | 70 | |
| XM_005266374.1 | XP_005266431.1 | 9 | 1104 | 70 | ||
| GenBank mRNA | AB208984.1 | - | 9 | 1051 | 70 | |
| AK223305.1 | - | 9 | 1141 | 70 | ||
| AK312393.1 | - | 9 | 1072 | 70 | ||
| BC007673.1 | - | 9 | 1044 | 3143290 | 70 | |
| BC010271.1 | - | 9 | 1108 | 2900752 | 70 | |
| CR936795.1 | - | 9 | 1094 | 70 | ||
| EU794648.1 | - | 9 | 1044 | 70 | ||
| J02923.1 | - | 9 | 1042 | 70 | ||
| M22300.1 | - | 7 | 797 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000642.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
T cell activation involved in immune response
protein kinase A signaling
cell migration
extracellular matrix disassembly
organ regeneration
regulation of intracellular protein transport
wound healing, spreading of cells
actin filament bundle assembly
positive regulation of podosome assembly
actin binding
calcium ion binding
identical protein binding
actin filament binding
GTPase binding
T cell activation involved in immune response
protein kinase A signaling
cell migration
extracellular matrix disassembly
organ regeneration
regulation of intracellular protein transport
wound healing, spreading of cells
actin filament bundle assembly
positive regulation of podosome assembly
actin binding
calcium ion binding
identical protein binding
actin filament binding
GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13007297.hg.1 | ENST00000323076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007297.hg.1 | ENST00000398576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007297.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007297.hg.1 | TC13000642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000416500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000442275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000460190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | ENST00000494531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008840.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000323076.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000398576.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | ENST00000469227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | NM_002298.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159655.hg.1 | TC1300008840.hg.1 | Clariom D TC |

LCP1
Genomic Map