Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
ENST00000603468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010301.hg.1 |
NR_015384.2 |
Refseq |