Assay Details
Gene Symbol: MIR610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030341.1 | 1 | 51 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin family member 18A | KIF18A |
MS-KIF18A, PPP1R99 |
Anti-sense | Chr. 11 - 28020616 - 28108199 on NCBI Build GRCh38 | |
Downstream | methyltransferase like 15 | METTL15 | METT5D1 | 51.3kb | Sense | Chr. 11 - 28108251 - 28527041 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007138.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000282.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114723 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114734 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714118 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714091 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114730 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114726 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714087 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114725 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114736 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114730 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114726 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114725 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114723 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714118 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114734 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714091 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714097 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114722 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114717 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28114708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07714087 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007138.hg.1 | ENST00000385139.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007138.hg.1 | NR_030341.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003725.hg.1 | ENST00000385139.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003725.hg.1 | NR_030341.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003725.hg.1 | TC11000282.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100150267.hg.1 | ENST00000385139.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150267.hg.1 | NR_030341.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100150267.hg.1 | ENST00000385139.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150267.hg.1 | NR_030341.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100150267.hg.1 | TC1100007138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007138.hg.1 | ENST00000385139.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007138.hg.1 | NR_030341.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309169 | NR_030341.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309169 | NR_030341.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309169 | NR_030341.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309169 | NR_030341.1 | human_transcriptome |
