Assay Details
Gene Symbol: MIR635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030365.1 | 1 | 48 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
WD repeat domain, phosphoinositide interacting 1 |
WIPI1 |
ATG18, ATG18A, WIPI49 |
Sense | Chr. 17 - 68421281 - 68457524 on NCBI Build GRCh38 | |
Downstream | family with sequence similarity 20 member A | FAM20A |
AI1G, AIGFS, FP2747 |
110.0kb | Sense | Chr. 17 - 68534561 - 68600954 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011562.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001823.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015314 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17011247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015332 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015322 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015322 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015332 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431842 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015314 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17011247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10431845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31015297 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17024385.hg.1 | ENST00000384830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17024385.hg.1 | NR_030365.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17024385.hg.1 | TC17001823.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011562.hg.1 | ENST00000384830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011562.hg.1 | NR_030365.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011562.hg.1 | ENST00000384830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011562.hg.1 | NR_030365.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700193548.hg.1 | ENST00000384830.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700193548.hg.1 | NR_030365.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700193548.hg.1 | ENST00000384830.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700193548.hg.1 | NR_030365.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700193548.hg.1 | TC1700011562.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308728 | NR_030365.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308728 | NR_030365.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308728 | NR_030365.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308728 | NR_030365.1 | human_transcriptome |
