Assay Details
Gene Symbol: MIR708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030598.1 | 1 | 46 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
asparaginyl-tRNA synthetase 2, mitochondrial (putative) |
NARS2 |
DFNB94, SLM5, asnRS |
-827.2kb | Sense | Chr. 11 - 78435961 - 78574864 on NCBI Build GRCh38 |
Downstream | microRNA 5579 | MIR5579 | 20.1kb | Sense | Chr. 11 - 79422169 - 79422226 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011758.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002142.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315903 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896474 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315910 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896470 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896476 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896477 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896507 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315905 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315902 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896473 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896472 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315906 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315881 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896474 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315921 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315915 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896470 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315902 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315906 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315904 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896477 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315905 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896473 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315881 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315903 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896476 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896507 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07896472 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28315910 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11025629.hg.1 | ENST00000390708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11025629.hg.1 | NR_030598.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11025629.hg.1 | TC11002142.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011758.hg.1 | ENST00000390708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011758.hg.1 | NR_030598.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011758.hg.1 | ENST00000390708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011758.hg.1 | NR_030598.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190933.hg.1 | ENST00000390708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190933.hg.1 | NR_030598.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190933.hg.1 | ENST00000390708.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190933.hg.1 | NR_030598.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190933.hg.1 | TC1100011758.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310875 | NR_030598.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310875 | NR_030598.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310875 | NR_030598.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310875 | NR_030598.1 | human_transcriptome |
