Assay Details
Gene Symbol: MIR1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031567.1 | 1 | 38 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Cdc42 guanine nucleotide exchange factor 9 | ARHGEF9 |
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2 |
-456.0 | Sense | Chr. X - 63634967 - 63786025 on NCBI Build GRCh38 |
Downstream | APC membrane recruitment protein 1 | AMER1 |
FAM123B, OSCS, WTX |
399.0kb | Sense | Chr. X - 64185117 - 64205744 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009871.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001094.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763124 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17352271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763105 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763116 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763092 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763119 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477524 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477493 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763090 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763131 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763094 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763127 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763107 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763125 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477509 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763088 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763113 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477509 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763094 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17352271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763104 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763106 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763117 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763092 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763090 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763119 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763105 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763127 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763108 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763109 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477524 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763131 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763107 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763089 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763125 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763088 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763124 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11763116 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32477493 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X014885.hg.1 | TC0X001094.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014885.hg.1 | ENST00000410600.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014885.hg.1 | NR_031567.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009871.hg.1 | ENST00000410600.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009871.hg.1 | NR_031567.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167549.hg.1 | ENST00000410600.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167549.hg.1 | NR_031567.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167549.hg.1 | ENST00000410600.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167549.hg.1 | NR_031567.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167549.hg.1 | TC0X00009871.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009871.hg.1 | ENST00000410600.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009871.hg.1 | NR_031567.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309274 | NR_031567.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309274 | NR_031567.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309274 | NR_031567.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309274 | NR_031567.1 | human_transcriptome |
