Assay Details
| Gene Symbol: MIR1286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031618.1 | 1 | 43 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 896 | LINC00896 | -40.6kb | Anti-sense | Chr. 22 - 20206332 - 20208537 on NCBI Build GRCh38 | |
| Downstream | proline dehydrogenase 1 pseudogene | LOC440792 | 48.1kb | Anti-sense | Chr. 22 - 20297310 - 20313590 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2200008029.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000509.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433154 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103885 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433154 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103922 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32103885 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11433153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008029.hg.1 | ENST00000408112.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008029.hg.1 | NR_031618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009425.hg.1 | ENST00000408112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009425.hg.1 | NR_031618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009425.hg.1 | TC22000509.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200157382.hg.1 | TC2200008029.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200157382.hg.1 | ENST00000408112.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157382.hg.1 | NR_031618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200157382.hg.1 | ENST00000408112.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157382.hg.1 | NR_031618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008029.hg.1 | ENST00000408112.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008029.hg.1 | NR_031618.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309909 | NR_031618.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309909 | NR_031618.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309909 | NR_031618.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309909 | NR_031618.1 | human_transcriptome |

MIR1286
Genomic Map