Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016865.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016865.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016865.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016865.hg.1 |
TC15001722.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016872.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016872.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016872.hg.1 |
TC15001722.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
ENST00000558578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
ENST00000561432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016869.hg.1 |
TC15001722.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185625.hg.1 |
TC1500010903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185639.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185639.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185639.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185639.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185639.hg.1 |
TC1500010903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000561432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000558913.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
ENST00000561432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
NR_033833.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500185634.hg.1 |
TC1500010903.hg.1 |
Clariom D TC |