Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030735.hg.1 |
TC01001984.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030735.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030735.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189884.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189884.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189884.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189884.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189884.hg.1 |
TC0100012210.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
ENST00000440494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012210.hg.1 |
NR_033883.1 |
Refseq |