Assay Details
| Gene Symbol: F11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033900.1 | 1 - 2 | 213 | 80 | ||
| GenBank mRNA | BC034307.1 | - | 1 - 2 | 154 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coagulation factor XI | F11 | FXI | -1.1kb | Anti-sense | Chr. 4 - 186265945 - 186290727 on NCBI Build GRCh38 |
| Downstream | melatonin receptor 1A | MTNR1A |
MEL-1A-R, MT1 |
31.9kb | Sense | Chr. 4 - 186532994 - 186555567 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002951.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04025046.hg.1 | TC04002951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025046.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025058.hg.1 | TC04002951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025058.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025058.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025058.hg.1 | NR_033901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025059.hg.1 | TC04002951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025059.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04025059.hg.1 | NR_033901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191548.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191548.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191548.hg.1 | TC0400013003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191547.hg.1 | TC0400013003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191549.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191549.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191549.hg.1 | TC0400013003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191527.hg.1 | TC0400013003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000505103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000508287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | ENST00000510172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400013003.hg.1 | NR_033900.1 | Refseq |

F11-AS1
Genomic Map