Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000641.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000641.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000641.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000641.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000638.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000638.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147253.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147253.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147253.hg.1 |
TC0600006914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147255.hg.1 |
TC0600006914.hg.1 |
Clariom D TC |