Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8116832 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8116834 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8123874 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8116833 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8123875 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8116831 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1560503_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000643.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000643.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000643.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000643.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000644.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000644.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000644.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000644.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000647.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000647.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000647.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000645.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000645.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000645.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000645.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000646.hg.1 |
TC06000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000646.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06000646.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147258.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147258.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147258.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147258.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147258.hg.1 |
TC0600006914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600147257.hg.1 |
TC0600006914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000420777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
ENST00000443546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600006914.hg.1 |
NR_033910.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1557070_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1560503_a_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557070_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1560503_PM_a_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557070_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1560503_a_at2 |
- |
- |
Human Genome U95 Set |
47125_PM_at |
- |
- |
Human Genome U95 Set |
47125_at |
- |
- |