| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010611.hg.1 |
TC09000821.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010611.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010612.hg.1 |
TC09000821.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010612.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010610.hg.1 |
TC09000821.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010610.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162922.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162922.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162922.hg.1 |
TC0900009233.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900162921.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162921.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162921.hg.1 |
TC0900009233.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900162923.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162923.hg.1 |
NR_033913.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900162923.hg.1 |
TC0900009233.hg.1 |
Clariom D TC |