Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022129.hg.1 |
TC04001588.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022129.hg.1 |
ENST00000512692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022129.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022130.hg.1 |
TC04001588.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022130.hg.1 |
ENST00000512692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022130.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022131.hg.1 |
TC04001588.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022131.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182084.hg.1 |
TC0400011969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011969.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000512692.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000608178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
ENST00000609616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
NR_033939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182085.hg.1 |
TC0400011969.hg.1 |
Clariom D TC |