Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014535.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014535.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014535.hg.1 |
TC12001100.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009614.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171866.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171866.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171866.hg.1 |
ENST00000536497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171866.hg.1 |
NR_033958.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171866.hg.1 |
TC1200009614.hg.1 |
Clariom D TC |