Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8104657 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8104656 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001253.hg.1 |
TC05000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001253.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147381.hg.1 |
ENST00000449530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147381.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147381.hg.1 |
ENST00000449530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147381.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147381.hg.1 |
TC0500007016.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500147383.hg.1 |
ENST00000412374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147383.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147383.hg.1 |
ENST00000412374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500147383.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147383.hg.1 |
TC0500007016.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500147384.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147384.hg.1 |
NR_033961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500147384.hg.1 |
TC0500007016.hg.1 |
Clariom D TC |