Assay Details
Gene Symbol: CASP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034065.2 | 3 - 4 | 692 | 121 | ||
NR_034066.2 | 2 - 3 | 440 | 121 | |||
NR_034067.2 | 3 - 4 | 692 | 121 | |||
NR_034068.2 | 2 - 3 | 440 | 121 | |||
NR_034071.2 | 4 - 5 | 787 | 121 | |||
GenBank mRNA | AF464194.1 | - | 3 - 4 | 692 | 121 | |
AF464195.1 | - | 2 - 3 | 440 | 121 | ||
AF486845.1 | - | 4 - 5 | 787 | 121 | ||
AF486846.1 | - | 3 - 4 | 692 | 121 | ||
AF486847.1 | - | 2 - 3 | 440 | 121 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102723895 | LOC102723895 | -281.1kb | Sense | Chr. 11 - 104568489 - 104609302 on NCBI Build GRCh38 | |
Downstream |
caspase 4, apoptosis-related cysteine peptidase pseudogene |
LOC643733 | 9.0kb | Sense | Chr. 11 - 104901549 - 104918175 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
