Assay Details
Gene Symbol: LINC01118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034099.1 | 1 - 2 | 252 | 62 | ||
GenBank mRNA | BC062774.1 | - | 1 - 2 | 241 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105374587 | LOC105374587 | -35.2kb | Sense | Chr. 2 - 46777785 - 46781473 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1119 | LINC01119 | 10.4kb | Sense | Chr. 2 - 46827864 - 46859006 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02004963.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02004367.hg.1 | TC02004963.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02004367.hg.1 | ENST00000409912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02004367.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02004366.hg.1 | TC02004963.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02004366.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02004366.hg.1 | ENST00000409912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02004366.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02004366.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151782.hg.1 | TC0200007482.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200151785.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151785.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151785.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151785.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151785.hg.1 | TC0200007482.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000468141.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000490950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | ENST00000495449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_024452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007482.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151784.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151784.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151784.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151784.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151784.hg.1 | TC0200007482.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000409518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000409912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000422294.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | ENST00000479311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | NR_034099.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200151783.hg.1 | TC0200007482.hg.1 | Clariom D TC |
