Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002510.hg.1 |
TC04000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002510.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002510.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148183.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148183.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148183.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148183.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148183.hg.1 |
TC0400007018.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148182.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148182.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148182.hg.1 |
ENST00000515882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400148182.hg.1 |
NR_037876.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400148182.hg.1 |
TC0400007018.hg.1 |
Clariom D TC |