Assay Details
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038219.1 | 1 - 2 | 130 | 66 | ||
GenBank mRNA | DA291077.1 | - | 1 - 2 | 118 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928231 | LOC101928231 | Anti-sense | Chr. 6 - 134429030 - 134478897 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC101928304 | LOC101928304 | 73.2kb | Anti-sense | Chr. 6 - 134525318 - 134540005 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001006.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06011858.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011858.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011852.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011852.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011852.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06011853.hg.1 | TC06001006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06011853.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165318.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165318.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165318.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600165317.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165317.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165317.hg.1 | NR_038218.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165317.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165317.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600165326.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165326.hg.1 | NR_038219.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600165326.hg.1 | TC0600009521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000262053.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000549612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000551831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | ENST00000552510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007622.hg.1 | NM_005171.4 | Refseq |
