Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021686.hg.1 |
TC02001382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021686.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021686.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021680.hg.1 |
TC02001382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021680.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021680.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021680.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021680.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
TC02001382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021681.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021685.hg.1 |
TC02001382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021685.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021685.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021685.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179217.hg.1 |
TC0200011041.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179213.hg.1 |
TC0200011041.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000434807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011041.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000447074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
ENST00000455128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179212.hg.1 |
TC0200011041.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200179218.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179218.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179218.hg.1 |
ENST00000415628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179218.hg.1 |
NR_038238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179218.hg.1 |
TC0200011041.hg.1 |
Clariom D TC |