Assay Details
Gene Symbol: LOC151484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038238.1 | 2 - 3 | 629 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | integral membrane protein 2C | ITM2C |
BRI3, BRICD2C, E25, E25C, ITM3 |
-16.1kb | Sense | Chr. 2 - 230864598 - 230879254 on NCBI Build GRCh38 |
Downstream | G protein-coupled receptor 55 | GPR55 | LPIR1 | 10.2kb | Anti-sense | Chr. 2 - 230907318 - 230961269 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001382.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083140 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956728 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956733 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083177 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083168 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956748 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083164 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083178 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956725 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956726 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083166 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083157 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22083138 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01956724 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 71202 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | AF367761.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | AL834229.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | BC033737.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | NM_001040102.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | NM_001040103.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128819 | NM_033207.3 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 721595 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 657973 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 827177 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 877794 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 499625 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 330030 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242631 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 151016 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3426943 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 41235 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P16073884 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P58135815 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3426943 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3426943 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P16073843 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00616381 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16073884 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16073843 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 110211 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 95691 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00616375 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13674011 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16073884 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16073843 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00616381 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00616383 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36109729 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00616386 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16073843 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16073884 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00616386 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00616381 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1562689_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000434807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000434807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02021689.hg.1 | TC02001382.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02021689.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02021687.hg.1 | TC02001382.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02021687.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02021687.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179219.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200179219.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179219.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200179219.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179219.hg.1 | TC0200011041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000415628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000434807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000434807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000447074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | ENST00000455128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011041.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179221.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179221.hg.1 | NR_038238.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200179221.hg.1 | TC0200011041.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1562689_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1562689_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562689_at2 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055170 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017078 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017078 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055170 | - | - |
