Assay Details
Gene Symbol: SOCS2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038263.1 | 3 - 4 | 457 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial ribosomal protein L42 | MRPL42 |
HSPC204, L31MT, L42MT, MRP-L31, MRP-L42, MRP-S32, MRPL31, MRPS32, PTD007, RPML31, S32MT |
-61.9kb | Anti-sense | Chr. 12 - 93467490 - 93503772 on NCBI Build GRCh38 |
Downstream | suppressor of cytokine signaling 2 | SOCS2 |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
2.9kb | Anti-sense | Chr. 12 - 93569822 - 93626236 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001817.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375002 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P333413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1559315_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR12023991.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023991.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12023991.hg.1 | TC12001817.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000547845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | ENST00000551626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | NR_038263.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200189441.hg.1 | TC1200011541.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1559315_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559315_s_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at11 | - | - |
Human Genome U95 Set | 64834_PM_at | - | - |
Human Genome U95 Set | 64834_at | - | - |
Human Genome U95 Set | 73954_PM_at | - | - |
Human Genome U95 Set | 73954_at | - | - |
Human Genome U95 Set | 70162_PM_at | - | - |
Human Genome U95 Set | 70162_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047893 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047893 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1699188 | AK054607.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
