Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027424.hg.1 |
TC02001698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027424.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027424.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027425.hg.1 |
TC02001698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027425.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027425.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188435.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188435.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188435.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188435.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188435.hg.1 |
TC0200012108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200188434.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188434.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188434.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188434.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188434.hg.1 |
TC0200012108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |