Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027423.hg.1 |
TC02001698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027423.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027423.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007089.hg.1 |
ENST00000423483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188431.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188431.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188431.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188431.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188431.hg.1 |
TC0200012108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200188432.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188432.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188432.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188432.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188432.hg.1 |
TC0200012108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007089.hg.1 |
ENST00000423483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188433.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188433.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188433.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188433.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188433.hg.1 |
TC0200012108.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
ENST00000418963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012108.hg.1 |
NR_038319.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1561370_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1561370_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561370_at7 |
- |
- |