Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006366.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006366.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006366.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006369.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006369.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006369.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006367.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006367.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006367.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006370.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006370.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006370.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006371.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006371.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006371.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006368.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006368.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006368.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400155353.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155353.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155353.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155353.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155353.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155356.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155356.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155356.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155356.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155356.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400155355.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155355.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155355.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155355.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155355.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400155357.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155357.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155357.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155357.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155357.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400155352.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155352.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155352.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155352.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155352.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400155354.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155354.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155354.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155354.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155354.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |