Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006373.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006373.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006373.hg.1 |
TC14000498.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007791.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155359.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155359.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155359.hg.1 |
ENST00000553979.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155359.hg.1 |
NR_038355.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155359.hg.1 |
TC1400007791.hg.1 |
Clariom D TC |