Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020194.hg.1 |
TC06001694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020194.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020194.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020188.hg.1 |
TC06001694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020188.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020188.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020188.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020193.hg.1 |
TC06001694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020193.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020193.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180462.hg.1 |
TC0600011729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600180469.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180469.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180469.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180469.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180469.hg.1 |
TC0600011729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000418399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
ENST00000453417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011729.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180471.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180471.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180471.hg.1 |
ENST00000416948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180471.hg.1 |
NR_038401.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180471.hg.1 |
TC0600011729.hg.1 |
Clariom D TC |