More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001370.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P52153 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10017344.hg.1 | ENST00000447119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017344.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017344.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017344.hg.1 | TC10001370.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010939.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017346.hg.1 | ENST00000447119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017346.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017346.hg.1 | TC10001370.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10017348.hg.1 | ENST00000447119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10017348.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10017348.hg.1 | TC10001370.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000174453.hg.1 | TC1000010939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000174455.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174455.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000174455.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000174455.hg.1 | TC1000010939.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000174455.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000447119.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | ENST00000449737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | NR_038453.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010939.hg.1 | NR_038453.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 239136_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 239136_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239136_at5 | - | - |
Human Genome U95 Set | 73577_PM_at | - | - |
Human Genome U95 Set | 73577_at | - | - |
Human Genome U133 Set | 239136_at | - | - |
Human Genome U133 Set | 239136_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042922 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042922 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 212060 | - | - |
