Assay Details
Gene Symbol: LOC285000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038891.1 | 1 - 2 | 235 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | four and a half LIM domains 2 | FHL2 |
AAG11, DRAL, FHL-2, SLIM-3, SLIM3 |
-162.2kb | Sense | Chr. 2 - 105360826 - 105438773 on NCBI Build GRCh38 |
Downstream | NCK adaptor protein 2 | NCK2 |
GRB4, NCKbeta |
134.4kb | Anti-sense | Chr. 2 - 105744649 - 105894274 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002168.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
