Assay Details
Gene Symbol: PRR7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038915.1 | 1 - 2 | 304 | 74 | ||
GenBank mRNA | BM083785.1 | - | 1 - 2 | 59 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05002106.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05029047.hg.1 | TC05002106.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029047.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029047.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029047.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029038.hg.1 | TC05002106.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029038.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029038.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029038.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029043.hg.1 | TC05002106.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029043.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029043.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029044.hg.1 | TC05002106.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029044.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029044.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029049.hg.1 | TC05002106.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029049.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029049.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190446.hg.1 | TC0500012973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500190449.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190449.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190449.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190449.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190449.hg.1 | TC0500012973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190436.hg.1 | TC0500012973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500190448.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190448.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190448.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190448.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190448.hg.1 | TC0500012973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000506465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012973.hg.1 | NR_038916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000511565.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190441.hg.1 | TC0500012973.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | ENST00000425316.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | ENST00000514846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | NR_038915.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500190442.hg.1 | TC0500012973.hg.1 | Clariom D TC |
