Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012181.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012181.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012181.hg.1 |
TC12000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012185.hg.1 |
TC12000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012184.hg.1 |
TC12000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012182.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012182.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012182.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012182.hg.1 |
TC12000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000620336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
ENST00000620336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000620336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
ENST00000620336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009117.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167355.hg.1 |
TC1200009117.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000535200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000538804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000539446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167354.hg.1 |
TC1200009117.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167350.hg.1 |
TC1200009117.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
ENST00000542314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
NR_038924.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167352.hg.1 |
TC1200009117.hg.1 |
Clariom D TC |